9,9-Dimethylfluorene-2,7-diboronic acid bis(pinacol) ester 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 9,9-Dimethylfluorene-2,7-diboronic acid Bis(pinacol) ester, 2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-dimethylfluorene, 2,2'-(9,9-Dimethyl-9H-fluorene-2,7-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)

H51954-03 H51954-06
AAH51954-03EA 63.9 USD
AAH51954-03 AAH51954-06
9,9-Dimethylfluorene-2,7-diboronic acid bis(pinacol) ester 95%
9,9-Dimethylfluorene-2,7-diboronic acid bis(pinacol) ester

Purity: 95% CAS No.: 325129-69-9 Molecular Formula: C27H36B2O4 Formula Weight: 446.20 MDL No.: MFCD08704230

Formula: C₂₇H₃₆B₂O₄
MW: 446.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08704230
CAS Number: 325129-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR