9,9-Dimethylfluorene-2-boronic acid pinacol ester 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 9,9-Dimethylfluorene-2-boronic acid Pinacol ester, 9,9-Dimethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl)-9H-fluorene, 2-(9,9-Dimethyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H51950-03 H51950-06
AAH51950-03EA 49.2 USD
AAH51950-03 AAH51950-06
9,9-Dimethylfluorene-2-boronic acid pinacol ester 95%
9,9-Dimethylfluorene-2-boronic acid pinacol ester

Purity: 95% CAS No.: 569343-09-5 Molecular Formula: C21H25BO2 Formula Weight: 320.24 MDL No.: MFCD08704229

Formula: C₂₁H₂₅BO₂
MW: 320.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08704229
CAS Number: 569343-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR