Fluorene-2-boronic acid pinacol ester 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
9H-Fluorene-2-boronic acid Pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolane-2-yl)-9H-fluorene, 2-(9H-Fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(9H-Fluoren-2-yl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane
H51947-03
H51947-06
AAH51947-03EA
38.6
USD
AAH51947-03
AAH51947-06
Fluorene-2-boronic acid pinacol ester 95%
Fluorene-2-boronic acid pinacol ester
Purity: 95%
Molecular Formula: C19H21BO2
Formula Weight: 292.18
MDL No.: MFCD16294548
Formula:
C₁₉H₂₁BO₂ MW: 292.18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16294548 CAS Number: 922706-40-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...