4-(Morpholino)phenylboronic acid pinacol ester 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine, 4-Morpholinophenylboronic acid Pinacol ester, 4-(4-Morpholinyl)benzeneboronic acid pinacol ester, 4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)morpholine
H51925-03
H51925-06
AAH51925-03EA
71.1
USD
AAH51925-03
AAH51925-06
4-(Morpholino)phenylboronic acid pinacol ester 95%
4-(Morpholino)phenylboronic acid pinacol ester
Purity: 95%
CAS No.: 568577-88-8
Molecular Formula: C16H24BNO3
Formula Weight: 289.18
MDL No.: MFCD04112544
Formula:
C₁₆H₂₄BNO₃ MW: 289.18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04112544 CAS Number: 568577-88-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...