4-(1-Piperidinyl)benzeneboronic acid pinacol ester 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine, 1-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperidine, 1-[4-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenyl]-piperidine
H51922-03
H51922-06
AAH51922-03EA
71.8
USD
AAH51922-03
AAH51922-06
4-(1-Piperidinyl)benzeneboronic acid pinacol ester 95%
4-(1-Piperidinyl)benzeneboronic acid pinacol ester
Purity: 95%
CAS No.: 852227-96-4
Molecular Formula: C17H26BNO2
Formula Weight: 287.21
MDL No.: MFCD07368524
Formula:
C₁₇H₂₆BNO₂ MW: 287.21 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07368524 CAS Number: 852227-96-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...