Anthracene-9,10-diboronic acid bis(pinacol) ester 95%

Supplier: Thermo Scientific Chemicals

H51921-06 H51921-03
AAH51921-06EA 258 USD
AAH51921-06 AAH51921-03
Anthracene-9,10-diboronic acid bis(pinacol) ester 95%
Anthracene-9,10-diboronic acid bis(pinacol) ester

Purity: 95% CAS No.: 863992-56-7 Molecular Formula: C26H32B2O4 Formula Weight: 430.16 MDL No.: MFCD16294538

Formula: C₂₆H₃₂B₂O₄
Storage Temperature: Ambient
MDL Number: MFCD16294538
CAS Number: 863992-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR