Benzene-1,4-dithiocarboxamide 97%

Supplier: Thermo Scientific Chemicals
Danger

H51857-03 H51857-06
AAH51857-03EA 69.4 USD
AAH51857-03 AAH51857-06
Benzene-1,4-dithiocarboxamide 97%
Benzene-1,4-dithiocarboxamide

Purity: 97% CAS No.: 13363-51-4 Molecular Formula: C8H8N2S2 Formula Weight: 196.28 UN No.: 2811 MDL No.: MFCD00046866

Formula: C₈H₈N₂S₂
MW: 196.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00046866
CAS Number: 13363-51-4
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR