2-Pyridin-3-yl-1H-benzimidazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H51785-06
H51785-14
AAH51785-06EA
65.27
USD
AAH51785-06
AAH51785-14
2-Pyridin-3-yl-1H-benzimidazole 97%
2-Pyridin-3-yl-1H-benzimidazole
Purity: 97%
CAS No.: 1137-67-3
Molecular Formula: C12H9N3
Formula Weight: 195.22
Odor: Odorless
Melting Point: 254-260°
MDL No.: MFCD00022665
Formula:
C₁₂H₉N₃ Melting Pt: 254…260 °C Storage Temperature: Ambient |
MDL Number:
MFCD00022665 CAS Number: 1137-67-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...