DSS (Disuccinimidyl suberate) 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Bis(2,5-dioxopyrrolidin-1-yl) octanedioate, Suberic acid bis(N-hydroxysuccinimide ester), Disuccinimidyl octanedioate, DSS
H51771-03
H51771-ME
AAH51771-03EA
139
USD
AAH51771-03
AAH51771-ME
DSS (Disuccinimidyl suberate) 97%
DSS (Disuccinimidyl suberate)
Purity: 97%
CAS No.: 68528-80-3
Molecular Formula: C16H20N2O8
Formula Weight: 368.34
Melting Point: 166-170°
MDL No.: MFCD00049059
Formula:
C₁₆H₂₀N₂O₈ MW: 368.34 g/mol Melting Pt: 166…170 °C Storage Temperature: Ambient |
MDL Number:
MFCD00049059 CAS Number: 68528-80-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...