4-Ethynylbenzeneboronic acid pinacol ester 95%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(4-Ethynyl-phenyl)-4,4,5,5-tetramethyl-[1,3,2]dioxaborolane, 2-(4-Ethynyl-phenyl)-4,4,5,5-tetramethyl-[1,3,2]-dioxaborolane

H51745-06 H51745-03
AAH51745-06EA 341 USD
AAH51745-06 AAH51745-03
4-Ethynylbenzeneboronic acid pinacol ester 95%
4-Ethynylbenzeneboronic acid pinacol ester

Purity: 95% CAS No.: 1034287-04-1 Molecular Formula: C14H17BO2 Formula Weight: 228.10 MDL No.: MFCD16294504

Formula: C₁₄H₁₇BO₂
Storage Temperature: Ambient
MDL Number: MFCD16294504
CAS Number: 1034287-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR