(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-tert-Butyl pyrrolidin-3-ylcarbamate, (3S)-(-)-3-(Boc-amino)pyrrolidine

H51729-06 H51729-14
AAH51729-06EA 141 USD
AAH51729-06 AAH51729-14
(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine 98%
(3S)-(-)-3-(tert-Butoxycarbonylamino)pyrrolidine

Purity: 98% CAS No.: 122536-76-9 Molecular Formula: C9H18N2O2 Formula Weight: 186.25 Melting Point: 50-54° Beilstein Reference: 5377813 Optical Rotation: -23° (c=1 in methanol) UN No.: 3259 MDL No.: MFCD00143194

Formula: C₉H₁₈N₂O₂
MW: 186.26 g/mol
Melting Pt: 50…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00143194
CAS Number: 122536-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR