Thiophene-2,5-diboronic acid bis(pinacol) ester 97%
Supplier: Thermo Scientific Chemicals
H51725-03
H51725-06
AAH51725-03EA
79.3
USD
AAH51725-03
AAH51725-06
Thiophene-2,5-diboronic acid bis(pinacol) ester 97%
Thiophene-2,5-diboronic acid bis(pinacol) ester
Purity: 97%
CAS No.: 175361-81-6
Molecular Formula: C16H26B2O4S
Formula Weight: 336.06
Melting Point: 216-220°
MDL No.: MFCD09800563
Formula:
C₁₆H₂₆B₂O₄S Melting Pt: 216…220 °C Storage Temperature: Ambient |
MDL Number:
MFCD09800563 CAS Number: 175361-81-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...