4-(Dimethylamino)pyridine 99%, prilled
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Dimethylaminopyridine, DMAP, N,N-Dimethylpyridin-4-amine
H51715-06
H51715-14
H51715-22
AAH51715-06EA
27
USD
AAH51715-06
AAH51715-14
AAH51715-22
4-(Dimethylamino)pyridine 99%, prilled
4-(Dimethylamino)pyridine
Purity: 99%
CAS No.: 1122-58-3
Molecular Formula: C7H10N2
Formula Weight: 122.17
Flash Point: 124°(255°F)
Melting Point: 110-113°
Boiling Point: 162°/50mm
Beilstein Reference: 110354
UN No.: 2928
MDL No.: MFCD00006418
Formula:
C₇H₁₀N₂ MW: 122.17 g/mol Boiling Pt: 195 Melting Pt: 110…113 °C Flash Pt: 124 °C (255 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006418 CAS Number: 1122-58-3 EINECS: 214-353-5 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...