1,4-Benzenediboronic Acid Bis(pinacol) Ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,4-Phenylenebis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane), 1,4-Bis(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 1,4-Phenylenediboronic acid, pinacol ester

H51707-03 H51707-06
AAH51707-03EA 58.7 USD
AAH51707-03 AAH51707-06
1,4-Benzenediboronic Acid Bis(pinacol) Ester 97%
1,4-Benzenediboronic acid bis(pinacol)ester

Purity: 97% CAS No.: 99770-93-1 Molecular Formula: C18H28B2O4 Formula Weight: 330.03 MDL No.: MFCD08276852

Formula: C₁₈H₂₈B₂O₄
Storage Temperature: Ambient
MDL Number: MFCD08276852
CAS Number: 99770-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR