8-Quinolineboronic acid pinacol ester 95%
Supplier: Thermo Scientific Chemicals
H51698-03
H51698-06
AAH51698-03EA
73.5
USD
AAH51698-03
AAH51698-06
8-Quinolineboronic acid pinacol ester 95%
8-Quinolineboronic acid pinacol ester
Purity: 95%
CAS No.: 190788-62-6
Molecular Formula: C15H18BNO2
Formula Weight: 255.12
MDL No.: MFCD05663894
Formula:
C₁₅H₁₈BNO₂ MW: 255.12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05663894 CAS Number: 190788-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...