Ethyl 5-aminovalerate hydrochloride 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 5-aminopentanoate hydrochloride
H51682-MD
H51682-03
AAH51682-MDEA
59.2
USD
AAH51682-MD
AAH51682-03
Ethyl 5-aminovalerate hydrochloride 96%
Ethyl-5-aminovalerate hydrochloride
Purity: 96%
CAS No.: 29840-57-1
Molecular Formula: C7H15NO2.HCl
Formula Weight: 181.66
MDL No.: MFCD12546205
Formula:
C₇H₁₆ClNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD12546205 CAS Number: 29840-57-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...