2-Hydroxy-5-pyridineboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-ol, 6-Hydroxypyridine-3-boronic acid pinacol ester

H51673-03 H51673-MD
AAH51673-03EA 73.4 USD
AAH51673-03 AAH51673-MD
2-Hydroxy-5-pyridineboronic acid pinacol ester 97%
2-Hydroxy-5-pyridineboronic acid pinacol ester

Purity: 97% CAS No.: 1054483-78-1 Molecular Formula: C11H16BNO3 Formula Weight: 221.06 MDL No.: MFCD06795671

Formula: C₁₁H₁₆BNO₃
MW: 221.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06795671
CAS Number: 1054483-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR