3-Methyl-2-(tributylstannyl)pyridine 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H51540-03
H51540-MD
AAH51540-03EA
267
USD
AAH51540-03
AAH51540-MD
3-Methyl-2-(tributylstannyl)pyridine 96%
3-Methyl-2-(tributylstannyl)pyridine
Purity: 96%
CAS No.: 259807-97-1
Molecular Formula: C18H33NSn
Formula Weight: 382.16
Storage Temperature: 2°C to 8°C
Formula:
C₁₈H₃₃NSn Storage Temperature: Ambient |
MDL Number:
MFCD07787404 CAS Number: 259807-97-1 UN: 2788 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...