5-Methyl-2-(tributylstannyl)thiazole 90+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H51531-03
H51531-MD
AAH51531-03EA
337
USD
AAH51531-03
AAH51531-MD
5-Methyl-2-(tributylstannyl)thiazole 90+%
5-Methyl-2-(tributylstannyl)thiazole
Purity: 90+%
CAS No.: 848613-91-2
Molecular Formula: C16H31NSSn
Formula Weight: 388.19
Storage Temperature: 2°C to 8°C
Formula:
C₁₆H₃₁NSSn MW: 388.19 Storage Temperature: Ambient |
MDL Number:
MFCD09025809 CAS Number: 848613-91-2 UN: 2788 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...