5-(Tributylstannyl)pyrimidine 96%

Supplier: Thermo Scientific Chemicals
Danger

H51458-03 H51458-MD
AAH51458-03EA 206 USD
AAH51458-03 AAH51458-MD
5-(Tributylstannyl)pyrimidine 96%
5-(Tributylstannyl)pyrimidine

Purity: 96% CAS No.: 144173-85-3 Molecular Formula: C16H30N2Sn Formula Weight: 369.13 UN No.: 2788 MDL No.: MFCD01319056

Formula: C₁₆H₃₀N₂Sn
MW: 369.13
Storage Temperature: Ambient
MDL Number: MFCD01319056
CAS Number: 144173-85-3
UN: 2788
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR