2-(Tributylstannyl)pyrazine 95%

Supplier: Thermo Scientific Chemicals
Danger

H51370-03 H51370-MD
AAH51370-03EA 139 USD
AAH51370-03 AAH51370-MD
2-(Tributylstannyl)pyrazine 95%
2-(Tributylstannyl)pyrazine

Purity: 95% CAS No.: 205371-27-3 Molecular Formula: C16H30N2Sn Formula Weight: 369.12 Flash Point: 110°(230°F) Refractive Index: 1.5160 Boiling Point: 130/1mm UN No.: 2788 MDL No.: MFCD01319077

Formula: C₁₆H₃₀N₂Sn
MW: 369.12
Boiling Pt: 130/1 mm
Density: 1.171
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01319077
CAS Number: 205371-27-3
UN: 2788
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR