tert-Butylmagnesium chloride 1 M in MeTHF (2-methyltetrahydrofuran)

Supplier: Thermo Scientific Chemicals
Danger

H51164-AE H51164-AP
AAH51164-AEEA 178 USD
AAH51164-AE AAH51164-AP
tert-Butylmagnesium chloride 1 M in MeTHF (2-methyltetrahydrofuran)
tert-Butylmagnesium chloride

CAS No.: 677-22-5 Molecular Formula: C4H9ClMg Formula Weight: 116.87 UN No.: 3399 MDL No.: MFCD00000465

Formula: C₄H₉ClMg
MW: 116.87 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000465
CAS Number: 677-22-5
UN: 3399
ADR: "4.3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR