3-Chlorophenylmagnesium bromide 1 M in MeTHF (2-methyltetrahydrofuran)

Supplier: Thermo Scientific Chemicals
Danger

H51161-AD H51161-AE
AAH51161-ADEA 252 USD
AAH51161-AD AAH51161-AE
3-Chlorophenylmagnesium bromide 1 M in MeTHF (2-methyltetrahydrofuran)
3-Chlorophenylmagnesium bromide

CAS No.: 36229-42-2 Molecular Formula: C6H4BrClMg Formula Weight: 215.76 UN No.: 2924 MDL No.: MFCD00672003

Formula: C₆H₄BrClMg
MW: 215.757 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00672003
CAS Number: 36229-42-2
UN: 2924
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR