1,3-Difluoroacetone 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H51145-03
H51145-06
AAH51145-03EA
104
USD
AAH51145-03
AAH51145-06
1,3-Difluoroacetone 98%
1,3-Difluoroacetone
Purity: 98%
CAS No.: 453-14-5
Molecular Formula: C3H4F2O
Formula Weight: 94.06
Boiling Point: 101-102°
UN No.: 2929
MDL No.: MFCD07368665
Formula:
C₃H₄F₂O MW: 94.06 g/mol Boiling Pt: 101…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD07368665 CAS Number: 453-14-5 UN: 2929 ADR: "6.1",II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...