3-Chloro-2-fluoroaniline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H51144-06
H51144-22
AAH51144-06EA
27.2
USD
AAH51144-06
AAH51144-22
3-Chloro-2-fluoroaniline 97%
3-Chloro-2-fluoroaniline
Purity: 97%
CAS No.: 2106-04-9
Molecular Formula: C6H5ClFN
Formula Weight: 145.56
Flash Point:
Formula:
C₆H₅ClFN Boiling Pt: 99 °C (14 -rr) Density: 1.324 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00069415 CAS Number: 2106-04-9 EINECS: 218-283-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...