N-(tert-Butoxycarbonyl)-L-tert-leucine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Boc-L-tert-leucine, Boc-Tle-OH, (S)-2-(Boc-amino)-3,3-dimethylbutyric Acid, (S)-2-(tert-Butoxycarbonylamino)-3,3-dimethylbutyric Acid
H51136-03
H51136-06
H51136-14
AAH51136-03EA
33.4
USD
AAH51136-03
AAH51136-06
AAH51136-14
N-(tert-Butoxycarbonyl)-L-tert-leucine 98%
N-(tert-Butoxycarbonyl)-L-tert-leucine
Purity: 98%
CAS No.: 62965-35-9
Molecular Formula: C11H21NO4
Formula Weight: 231.29
Melting Point: 118-121°
MDL No.: MFCD00065574
Formula:
C₁₁H₂₁NO₄ MW: 231.29 g/mol Melting Pt: 118…121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00065574 CAS Number: 62965-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...