2-(Tributylstannyl)thiazole 96%

Supplier: Thermo Scientific Chemicals
Danger

H51090-03 H51090-06
AAH51090-03EA 135 USD
AAH51090-03 AAH51090-06
2-(Tributylstannyl)thiazole 96%
2-(Tributylstannyl)thiazole

Purity: 96% CAS No.: 121359-48-6 Molecular Formula: C15H29NSSn Formula Weight: 374.17 Boiling Point: 307-309° UN No.: 2788 MDL No.: MFCD01319057

Formula: C₁₅H₂₉NSSn
MW: 374.17
Boiling Pt: 307…309 °C
Density: 1.19
Storage Temperature: Ambient
MDL Number: MFCD01319057
CAS Number: 121359-48-6
UN: 2788
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR