2-Cyano-6-methoxybenzothiazole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Methoxy-2-benzothiazolecarbonitrile, 6-Methoxybenzo[d]thiazole-2-carbonitrile
H51082-03
H51082-06
AAH51082-03EA
107
USD
AAH51082-03
AAH51082-06
2-Cyano-6-methoxybenzothiazole 99%
2-Cyano-6-methoxybenzothiazole
Purity: 99%
CAS No.: 943-03-3
Molecular Formula: C9H6N2OS
Formula Weight: 190.22
MDL No.: MFCD00010537
Formula:
C₉H₆N₂OS MW: 190.22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010537 CAS Number: 943-03-3 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...