2,5-Bis(3,7-dimethyloctyloxy)terephthalaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H51067-03
H51067-MD
AAH51067-03EA
133
USD
AAH51067-03
AAH51067-MD
2,5-Bis(3,7-dimethyloctyloxy)terephthalaldehyde 98%
2,5-Bis(3,7-dimethyloctyloxy)terephthalaldehyde
Purity: 98%
CAS No.: 325461-35-6
Molecular Formula: C28H46O4
Formula Weight: 446.66
MDL No.: MFCD03427237
Formula:
C₂₈H₄₆O₄ MW: 446.66 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03427237 CAS Number: 325461-35-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...