1-Aza-18-crown-6 (1,4,7,10,13-pentaoxa-16-azacyclooctadecane) 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,4,7,10,13-Pentaoxa-16-azacyclooctadecane, 1-Aza-18-crown 6-Ether, 1-Aza-18-crown-6
H51065-03
H51065-MD
H51065-06
AAH51065-03EA
245
USD
AAH51065-03
AAH51065-MD
AAH51065-06
1-Aza-18-crown-6 (1,4,7,10,13-pentaoxa-16-azacyclooctadecane) 95%
1-Aza-18-crown-6 (1,4,7,10,13-Pentaoxa-16-azacyclooctadecane)
Purity: 95%
CAS No.: 33941-15-0
Molecular Formula: C12H25NO5
Formula Weight: 263.33
Melting Point: 46-49°
Beilstein Reference: 1074079
Formula:
C₁₂H₂₅NO₅ MW: 263.33 g/mol Melting Pt: 46…49 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075466 CAS Number: 33941-15-0 EINECS: 251-751-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...