1-Aza-15-crown-5 (1,4,7,10-tetraoxa-13-azacyclopentadecane) 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,4,7,10-Tetraoxa-13-azacyclopentadecane, 1-Aza-15-crown 5-Ether, 1,4,7,10-Tetraoxa-13-azacyclopentadecane

H51064-03 H51064-MD H51064-06
AAH51064-03EA 188 USD
AAH51064-03 AAH51064-MD AAH51064-06
1-Aza-15-crown-5 (1,4,7,10-tetraoxa-13-azacyclopentadecane) 97%
1-Aza-15-crown-5 (1,4,7,10-tetraoxa-13-azacyclopentadecane)

Purity: 97% CAS No.: 66943-05-3 Molecular Formula: C10H21NO4 Formula Weight: 219.28 Beilstein Reference: 971959 MDL No.: MFCD00075465

Formula: C₁₀H₂₁NO₄
MW: 219.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00075465
CAS Number: 66943-05-3
EINECS: 266-523-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR