1,10-Diaza-18-crown-6 (1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22) 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Diaza-18-crown-6, 1,4,10,13-Tetraoxa-7,16-diazacyclooctadecane
H51063-03
H51063-06
H51063-MD
AAH51063-03EA
177
USD
AAH51063-03
AAH51063-06
AAH51063-MD
1,10-Diaza-18-crown-6 (1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22) 96%
1,10-Diaza-18-crown-6 (1,4,10,13-tetraoxa-7,16-diazacyclooctadecane kryptofix® 22)
Purity: 96%
CAS No.: 23978-55-4
Molecular Formula: C12H26N2O4
Formula Weight: 262.35
Beilstein Reference: 609764
MDL No.: MFCD00005112
Formula:
C₁₂H₂₆N₂O₄ MW: 262.35 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00005112 CAS Number: 23978-55-4 EINECS: 245-965-0 UN: 3259 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...