1-Chloro-4-ethynylbenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(4-Chlorophenyl)acetylene, 1-Ethynyl-4-chlorobenzene, (4-Chlorophenyl)ethyne
H51058-03
H51058-06
AAH51058-03EA
169
USD
AAH51058-03
AAH51058-06
1-Chloro-4-ethynylbenzene 98%
1-Chloro-4-ethynylbenzene
Purity: 98%
CAS No.: 873-73-4
Molecular Formula: C8H5Cl
Formula Weight: 136.58
Flash Point: 57°(135°F)
Melting Point: 43-47°
Boiling Point: 179°
UN No.: 1325
MDL No.: MFCD00191917
Formula:
C₈H₅Cl MW: 136.58 g/mol Boiling Pt: 179 °C Melting Pt: 43…47 °C Flash Pt: 57 °C (135 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00191917 CAS Number: 873-73-4 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...