tert-Butyl propiolate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Propynoic acid tert-butyl ester, Propiolic acid tert-butyl ester

H51048-03 H51048-06
AAH51048-03EA 141 USD
AAH51048-03 AAH51048-06
tert-Butyl propiolate 98%
tert-Butyl propiolate

Purity: 98% CAS No.: 13831-03-3 Molecular Formula: C7H10O2 Formula Weight: 126.16 Flash Point: 28°(82°F) Refractive Index: 1.4180 Melting Point: 18-20° Boiling Point: 52-53°/27mm Beilstein Reference: 1747175 UN No.: 3272 MDL No.: MFCD00060100

Formula: C₇H₁₀O₂
MW: 126.16 g/mol
Boiling Pt: 52…53 °C/27 mm
Melting Pt: 18…20 °C
Density: 0.919
Flash Pt: 28 °C(82 °F)
Storage Temperature: Ambient
MDL Number: MFCD00060100
CAS Number: 13831-03-3
UN: 3272
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR