2-Chloro-5,5-dimethyl-1,3-cyclohexanedione 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H51035-03
H51035-MD
AAH51035-03EA
200
USD
AAH51035-03
AAH51035-06
AAH51035-MD
2-Chloro-5,5-dimethyl-1,3-cyclohexanedione 98%
2-Chloro-5,5-dimethyl-1,3-cyclohexanedione
Purity: 98%
CAS No.: 7298-89-7
Molecular Formula: C8H11ClO2
Formula Weight: 174.62
Melting Point: 161-164°
Beilstein Reference: 2326749
MDL No.: MFCD00010502
Formula:
C₈H₁₁ClO₂ Melting Pt: 161…164 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010502 CAS Number: 7298-89-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...