4-(Dimethylamino)butylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N1,N1-Dimethylbutane-1,4-diamine

H50276-03 H50276-06
AAH50276-03EA 107 USD
AAH50276-03 AAH50276-06
4-(Dimethylamino)butylamine 98%
4-(Dimethylamino)butylamine

Purity: 98% CAS No.: 3529-10-0 Molecular Formula: C6H16N2 Formula Weight: 116.21 Refractive Index: 1.4400 UN No.: 2735 MDL No.: MFCD00047977

Formula: C₆H₁₆N₂
MW: 116.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047977
CAS Number: 3529-10-0
EINECS: 222-559-1
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR