4-(Trifluoromethyl)benzoic anhydride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H34480-03
H34480-06
AAH34480-03EA
32.5
USD
AAH34480-03
AAH34480-06
4-(Trifluoromethyl)benzoic anhydride 97%
4-(Trifluoromethyl)benzoic anhydride
Purity: 97%
CAS No.: 25753-16-6
Molecular Formula: C16H8F6O3
Formula Weight: 362.23
Melting Point: 127-131°
MDL No.: MFCD00671577
Formula:
C₁₆H₈F₆O₃ MW: 362.23 g/mol Melting Pt: 127…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD00671577 CAS Number: 25753-16-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...