2-Bromo-6-iodopyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H34478-03
H34478-06
AAH34478-03EA
83.2
USD
AAH34478-03
AAH34478-06
AAH34478-MD
2-Bromo-6-iodopyridine 97%
2-Bromo-6-iodopyridine
Purity: 97%
CAS No.: 234111-08-1
Molecular Formula: C5H3BrIN
Formula Weight: 283.89
Melting Point: 139-143°
MDL No.: MFCD08059557
Formula:
C₅H₃BrIN Melting Pt: 139…143 °C Storage Temperature: Ambient |
MDL Number:
MFCD08059557 CAS Number: 234111-08-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...