3-Fluoro-5-iodoaniline 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H34457-03
H34457-06
AAH34457-03EA
113
USD
AAH34457-03
AAH34457-06
AAH34457-MD
3-Fluoro-5-iodoaniline 96%
3-Fluoro-5-iodoaniline
Purity: 96%
CAS No.: 660-49-1
Molecular Formula: C6H5FIN
Formula Weight: 237.02
MDL No.: MFCD04116466
Formula:
C₆H₅FIN Storage Temperature: Ambient |
MDL Number:
MFCD04116466 CAS Number: 660-49-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...