2-Formyl-6-methoxypyridine 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Methoxypicolinaldehyde, 6-Methoxypyridine-2-carboxaldehyde, 6-Methoxypyridine-2-carbaldehyde

H34432-03 H34432-06
AAH34432-03EA 66.4 USD
AAH34432-03 AAH34432-06
2-Formyl-6-methoxypyridine 96%
2-Formyl-6-methoxypyridine

Purity: 96% CAS No.: 54221-96-4 Molecular Formula: C7H7NO2 Formula Weight: 137.14 Refractive Index: 1.5310

Formula: C₇H₇NO₂
MW: 137.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05664030
CAS Number: 54221-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR