4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(4-Fluorobenzoyl)-1,1,1-trifluoroacetone
H34429-03
H34429-06
H34429-14
AAH34429-03EA
16.3
USD
AAH34429-03
AAH34429-06
AAH34429-14
4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione 97%
4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione
Purity: 97%
CAS No.: 582-65-0
Molecular Formula: C10H6F4O2
Formula Weight: 234.15
Melting Point: 40-44°
MDL No.: MFCD03419766
Formula:
C₁₀H₆F₄O₂ MW: 234.15 g/mol Melting Pt: 40…44 °C Storage Temperature: Ambient |
MDL Number:
MFCD03419766 CAS Number: 582-65-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...