3-(2-Bromophenyl)-1H-pyrazole 97%

Supplier: Thermo Scientific Chemicals

H34406-03 H34406-09
AAH34406-03EA 40.6 USD
AAH34406-03 AAH34406-09
3-(2-Bromophenyl)-1H-pyrazole 97%
3-(2-Bromophenyl)-1H-pyrazole

Purity: 97% CAS No.: 114382-20-6 Molecular Formula: C9H7BrN2 Formula Weight: 223.07 Melting Point: 130-132° MDL No.: MFCD01940435

Formula: C₉H₇BrN₂
MW: 223.07 g/mol
Melting Pt: 130…132 °C
Storage Temperature: Ambient
MDL Number: MFCD01940435
CAS Number: 114382-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR