Furo[3,2-b]pyridine-2-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H34400-03 H34400-06 H34400-MD
AAH34400-03EA 143 USD
AAH34400-03 AAH34400-06 AAH34400-MD
Furo[3,2-b]pyridine-2-carboxylic acid 97%
Furo[3,2-b]pyridine-2-carboxylic acid

Purity: 97% CAS No.: 112372-14-2 Molecular Formula: C8H5NO3 Formula Weight: 163.13 Melting Point: 300° MDL No.: MFCD11588128

Formula: C₈H₅NO₃
MW: 163.13 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD11588128
CAS Number: 112372-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR