3-(m-Tolyl)propionic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(3-Methylphenyl)propionic acid, 3-(3-Methylphenyl)propanoic acid

H34395-03 H34395-06
AAH34395-03EA 73.2 USD
AAH34395-03 AAH34395-06
3-(m-Tolyl)propionic acid 96%
3-(m-Tolyl)propionic acid

Purity: 96% CAS No.: 3751-48-2 Molecular Formula: C10H12O2 Formula Weight: 164.20 Melting Point: 42-44° MDL No.: MFCD00060326

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Melting Pt: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00060326
CAS Number: 3751-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR