trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H34384-14
H34384-22
AAH34384-06EA
26.85
USD
AAH34384-06
AAH34384-14
AAH34384-22
trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid 98%
trans-4-(4-Chlorophenyl)cyclohexane-1-carboxylic acid
Purity: 98%
CAS No.: 49708-81-8
Molecular Formula: C13H15ClO2
Formula Weight: 238.71
Melting Point: 249-254°
MDL No.: MFCD06797626
Formula:
C₁₃H₁₅ClO₂ Melting Pt: 249…254 °C Storage Temperature: Ambient |
MDL Number:
MFCD06797626 CAS Number: 49708-81-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...