3-(o-Tolyl)propionic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(2-Methylphenyl)propionic acid, 2-Methylhydrocinnamic acid, 3-(2-Methylphenyl)propanoic acid

H34375-03 H34375-09
AAH34375-03EA 37.2 USD
AAH34375-03 AAH34375-09
3-(o-Tolyl)propionic acid 96%
3-(o-Tolyl)propionic acid

Purity: 96% CAS No.: 22084-89-5 Molecular Formula: C10H12O2 Formula Weight: 164.20 Melting Point: 101-103° MDL No.: MFCD00079773

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Melting Pt: 101…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00079773
CAS Number: 22084-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR