Methyl-4-amino-3-nitrobenzoate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 4-amino-3-nitrobenzoate
H34348-03
H34348-09
AAH34348-03EA
33.2
USD
AAH34348-03
AAH34348-09
Methyl-4-amino-3-nitrobenzoate 97%
Methyl 4-amino-3-nitrobenzoate
Purity: 97%
CAS No.: 3987-92-6
Molecular Formula: C8H8N2O4
Formula Weight: 196.16
Melting Point: 188-191°
MDL No.: MFCD00017562
Formula:
C₈H₈N₂O₄ MW: 196.16 g/mol Melting Pt: 188…191 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017562 CAS Number: 3987-92-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...