3-(4-Chloro-3-fluorophenyl)propionic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

H34205-03 H34205-06
AAH34205-03EA 39.1 USD
AAH34205-03 AAH34205-06
3-(4-Chloro-3-fluorophenyl)propionic acid 96%
3-(4-Chloro-3-fluorophenyl)propionic acid

Purity: 96% CAS No.: 881189-65-7 Molecular Formula: C9H8ClFO2 Formula Weight: 202.61 Melting Point: 80-84° MDL No.: MFCD04116058

Formula: C₉H₈ClFO₂
MW: 202.61 g/mol
Melting Pt: 80…84 °C
Storage Temperature: Ambient
MDL Number: MFCD04116058
CAS Number: 881189-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR