3-Bromo-4-iodopyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H34203-03
H34203-MD
AAH34203-03EA
133
USD
AAH34203-03
AAH34203-MD
3-Bromo-4-iodopyridine 97%
3-Bromo-4-iodopyridine
Purity: 97%
CAS No.: 89167-19-1
Molecular Formula: C5H3BrIN
Formula Weight: 283.89
Melting Point: 111-115°
MDL No.: MFCD11110263
Formula:
C₅H₃BrIN Melting Pt: 111…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD11110263 CAS Number: 89167-19-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...