2-Amino-5-iodonicotinonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H34191-03
H34191-06
AAH34191-03EA
84.7
USD
AAH34191-03
AAH34191-06
AAH34191-MD
2-Amino-5-iodonicotinonitrile 97%
2-Amino-5-iodonicotinonitrile
Purity: 97%
CAS No.: 1347815-41-1
Molecular Formula: C6H4IN3
Formula Weight: 245.02
Melting Point: 193-197°
UN No.: 3439
MDL No.: MFCD20040120
Formula:
C₆H₄IN₃ Melting Pt: 193…197 °C Storage Temperature: Ambient |
MDL Number:
MFCD20040120 CAS Number: 1347815-41-1 UN: 3439 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...